Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2C/CD159c/KLRC2 Antibody (2098A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB349965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349965 Supplier Novus Biologicals Supplier No. FAB1382MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NKG2C/CD159c/KLRC2 Monoclonal antibody specifically detects NKG2C/CD159c/KLRC2 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen NKG2C/CD159c/KLRC2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2098A
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor
Host Species Rabbit
Immunogen Mouse NKG2C/CD159c synthetic peptide, Accession # NP_034783
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3822
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.