Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2D/CD314 Antibody (AP-MAB0840), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB257485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB257485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB257485 Supplier Novus Biologicals Supplier No. NBP106652AF350
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen NKG2D/CD314
Applications Flow Cytometry
Classification Monoclonal
Clone AP-MAB0840
Conjugate Alexa Fluor 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
Host Species Armenian Hamster
Immunogen Soluble mouse NKG2D.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 22914
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.