Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2E/KLRC3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB326855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326855 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326855 Supplier Novus Biologicals Supplier No. NBP299155PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKG2E/KLRC3 Polyclonal antibody specifically detects NKG2E/KLRC3 in Human samples. It is validated for Western Blot

Specifications

Antigen NKG2E/KLRC3
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias killer cell lectin-like receptor subfamily C, member 3, NK cell receptor E, NKG2-E type II integral membrane protein, NKG2-E-activating NK receptor, NKG2ENKG2-E
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NKG2E/KLRC3
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3823
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.