Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIAMRE Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018107 0.025 mg
NB200123 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018107 Supplier Novus Biologicals Supplier No. NB200123SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKIAMRE Polyclonal specifically detects NKIAMRE in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen NKIAMRE
Applications Immunoprecipitation, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias cyclin-dependent kinase-like 3, EC 2.7.11, EC 2.7.11.22, NKIAMRESerine/threonine-protein kinase NKIAMRE, serine-threonine protein kinase NKIAMRE
Gene Symbols CDKL3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 414-428 [NCNGLKENPHCGGSV] of the 50 kDa human NKIAMRE protein.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51265
Test Specificity Reacts with residues 414-428 [NCNGLKENPHCGGSV] of the 50 kDa human NKIAMRE protein.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.