Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIRAS1 Antibody (184C278), HRP, Novus Biologicals™
SDP

Catalog No. NB10056555H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056555H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056555H Supplier Novus Biologicals Supplier No. NB10056555H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKIRAS1 Monoclonal specifically detects NKIRAS1 in Bovine, Canine, Equine, Human, Mouse, Pig, Primate, Rat, Xenopus species samples. It is validated for Western Blot.

Specifications

Antigen NKIRAS1
Applications Western Blot
Classification Monoclonal
Clone 184C278
Conjugate HRP
Dilution Western Blot
Gene Accession No. Q9NYS0
Gene Alias I-kappa-B-interacting Ras-like protein 1, kappa B-ras 1, Kappa B-Ras protein 1, KappaB-Ras1, KBRAS1kappaB-Ras1, NF-kappa-B inhibitor-interacting Ras-like protein 1, NFKB inhibitor interacting Ras-like 1, NFKB inhibitor interacting Ras-like protein 1
Gene Symbols NKIRAS1
Host Species Mouse
Immunogen This monoclonal antibody was raised against a synthetic peptide containing amino acid residues 96-113 (RVELLKKEIDKFKDKKEV) of human kB-Ras1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28512
Target Species Bovine, Canine, Equine, Human, Mouse, Pig, Primate, Rat, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.