Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp30/NCR3 Antibody (210845), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB367285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367285 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367285 Supplier Novus Biologicals Supplier No. FAB1849Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Agonist Activity, CyTOF, Flow Cytometry.

Specifications

Antigen NKp30/NCR3
Applications Agonist Activity, CyTOF, Flow Cytometry
Classification Monoclonal
Clone 210845
Conjugate DyLight 680
Dilution Flow Cytometry, Agonist Activity, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS
Host Species Mouse
Immunogen Mouse T cell hybridoma transfected with human NKp30/NCR3 and Mouse myeloma cell line NS0-derived recombinant human NKp30 Fc Chimera
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 259197
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.