Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp30/NCR3 Antibody (2615A), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB367305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367305 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367305 Supplier Novus Biologicals Supplier No. FAB18492K
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NKp30/NCR3 Monoclonal specifically detects NKp30/NCR3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen NKp30/NCR3
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 2615A
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Activating natural killer receptor p30, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NKP30, NKp30MALS
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human NKp30/NCR3, Leu19-Thr138, Accession # Q05D23
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Checkpoint signaling, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 259197
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.