Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp44/NCR2 Antibody (1G6), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N242683APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N242683APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N242683APC7 Supplier Novus Biologicals Supplier No. NBP242683APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKp44/NCR2 Monoclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NKp44/NCR2
Applications Flow Cytometry
Classification Monoclonal
Clone 1G6
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD336, CD336 antigen, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1
Gene Symbols NCR2
Host Species Mouse
Immunogen Recombinant human NKp44 (19-130aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9436.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.