Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp46/NCR1 Antibody (B-L46), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB388338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388338 Supplier Novus Biologicals Supplier No. NBP314597APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKp46/NCR1 Monoclonal antibody specifically detects NKp46/NCR1 in Human samples. It is validated for Flow Cytometry, Functional Assay

Specifications

Antigen NKp46/NCR1
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone B-L46
Conjugate APC
Formulation PBS
Gene Alias CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Host Species Mouse
Immunogen Recombinant human NKp46/NCR1
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9437
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.