Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp46/NCR1 Antibody (MM0491-8F24) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211820 0.1 mg
NB018910 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211820 Supplier Novus Biologicals Supplier No. NBP211820

Mouse Monoclonal Antibody

NKp46/NCR1 Monoclonal specifically detects NKp46/NCR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen NKp46/NCR1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MM0491-8F24
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:20-1:100, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. AAH64806
Gene Alias CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46)
Gene Symbols NCR1
Host Species Mouse
Immunogen Human recombinant NKp46/NCR1
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 9437
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.