Learn More
Description
Specifications
Specifications
| Antigen | NKp46/NCR1 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | n1D9 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Gene Accession No. | NP_004820 |
| Gene Alias | CD335, CD335 antigen, hNKp46, LY94lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46), Lymphocyte antigen 94 homolog, natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein, NK cell-activating receptor, NKp46, NKP46FLJ99094, NK-p46lymphocyte antigen 94 (mouse) homolog (activating NK-receptor; NK-p46) |
| Gene Symbols | NCR1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
