Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp80/KLRF1 Antibody (239127), HRP, Novus Biologicals™
SDP

Catalog No. NB296951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296951 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296951 Supplier Novus Biologicals Supplier No. FAB1900H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

NKp80/KLRF1 Monoclonal specifically detects NKp80/KLRF1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NKp80/KLRF1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 239127
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human NKp80/KLRF1, Val66-Tyr231, Accession # Q9NZS2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51348
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.