Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp80/KLRF1 Antibody (MM0492-7D32), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N211821PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N211821PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N211821PEC5 Supplier Novus Biologicals Supplier No. NBP211821PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

NKp80/KLRF1 Monoclonal antibody specifically detects NKp80/KLRF1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen NKp80/KLRF1
Applications Flow Cytometry
Classification Monoclonal
Clone MM0492-7D32
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Activating coreceptor NKp80, CLEC5CMGC119908, C-type lectin domain family 5 member C, killer cell lectin-like receptor F1, killer cell lectin-like receptor subfamily F member 1, killer cell lectin-like receptor subfamily F, member 1, Lectin-like receptor F1, MGC119907, MGC119909, ML, NKp80
Gene Symbols KLRF1
Host Species Rat
Immunogen Human recombinant NKp80/KLRF1
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51348.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.