Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKR-P1 Antibody (3.2.3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222437 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 ug
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222437 500 μg
NB437691 100 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222437 Supplier Novus Biologicals Supplier No. NBP222437

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

NKR-P1 Monoclonal specifically detects NKR-P1 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen NKR-P1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 3.2.3
Concentration 0.51 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready
Gene Alias killer cell lectin-like receptor subfamily B, member 1, Klrb1, Klrb1 killer cell lectin-like receptor subfamily B, member 1, Klrb1d, Klrb1f, Klrb1g, Klrb6, Nkr-p1e, Nkrp-1e, NKR-P1G
Gene Symbols KLRB1
Host Species Mouse
Immunogen Rat 24.5 kD cell surface protein known as NKR-P1
Purification Method Protein G purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 689817
Test Specificity This antibody reacts with Rat.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.