Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKR-P1 Antibody (3.2.3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | NKR-P1 |
---|---|
Clone | 3.2.3 |
Concentration | 0.51 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
Description
NKR-P1 Monoclonal specifically detects NKR-P1 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
NKR-P1 | |
0.51 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Rat | |
689817 | |
Rat 24.5 kD cell surface protein known as NKR-P1 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
3.2.3 | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
killer cell lectin-like receptor subfamily B, member 1, Klrb1, Klrb1 killer cell lectin-like receptor subfamily B, member 1, Klrb1d, Klrb1f, Klrb1g, Klrb6, Nkr-p1e, Nkrp-1e, NKR-P1G | |
KLRB1 | |
IgG1 κ | |
Protein G purified | |
This antibody reacts with Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title