Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKR-P1B (CD161b) Mouse anti-Mouse, Unconjugated, Clone: 2D12, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $481.52
Specifications
Antigen | NKR-P1B (CD161b) |
---|---|
Clone | 2D12 |
Applications | Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21152MI
|
MilliporeSigma
MABF211525UL |
25 μL |
Each for $189.29
|
|
|||||
MABF21151MI
|
MilliporeSigma
MABF2115100UL |
100 μL |
Each for $481.52
|
|
|||||
Description
NKR-P1B (CD161b) Monoclonal antibody specifically detects NKR-P1B (CD161b) in Mouse samples. It is validated for Flow Cytometry, Function Assay, Immunoprecipitation, Western Blotting.Specifications
NKR-P1B (CD161b) | |
Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Mouse | |
Killer cell lectin-like receptor subfamily B member 1B allele B;CD161 antigen-like family member B;Inhibitory receptor NKR-P1B;Lymphocyte antigen 55d;Ly-55d;Natural killer cell surface protein NKR-P1B allele B6;CD161b | |
Mouse BaF/3 cells (murine IL-3-dependent pro-B cell line) expressing mouse NKR-P1B protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼52 kDa observed; 25.03 kDa calculated. . Uncharacterized bands may be observed in some lysate(s). |
2D12 | |
Monoclonal | |
Purified | |
Inflammation and Immunology | |
NP_085102.4 | |
Klrb1b, Klrb1d, Ly55d, Nkrp1b, Nkrp1d | |
IgG2a λ | |
Protein G Purified | |
Clone 2D12 is a mouse monoclonal antibody that detects Natural killer cell surface protein NKR-P1B allele B6 (NKR-P1B) in murine species. It does not react with Nkrp1a, Nkrp1c, and Nkrp1f. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title