Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKR-P1D Antibody (4E9), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250531AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250531AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250531AH Supplier Novus Biologicals Supplier No. NBP250531AF647

Rat Monoclonal Antibody

NKR-P1D Monoclonal specifically detects NKR-P1D in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen NKR-P1D
Applications Flow Cytometry
Classification Monoclonal
Clone 4000000000
Conjugate Alexa Fluor 647
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Rat
Immunogen YB2/0 rat plasmacytoma cells transfected with NKRP1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.