Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKRF Antibody (OTI4C2), Biotin, Novus Biologicals™
SDP

Catalog No. NBP273001B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273001B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273001B Supplier Novus Biologicals Supplier No. NBP273001B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKRF Monoclonal specifically detects NKRF in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen NKRF
Applications Western Blot
Classification Monoclonal
Clone OTI4C2
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias ITBA4NFkB-repressing factor, NF-kappaB repressing factor, NFKB repressing factor, NRFNF-kappa-B-repressing factor, Protein ITBA4, Transcription factor NRF
Gene Symbols NKRF
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-296 of human NKRF (NP_060014) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55922.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.