Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKRF Antibody (OTI4C2), Novus Biologicals™
SDP

Catalog No. NBP246173 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246173 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246173 Supplier Novus Biologicals Supplier No. NBP246173
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

NKRF Monoclonal specifically detects NKRF in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen NKRF
Applications Western Blot
Classification Monoclonal
Clone OTI4C2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias ITBA4NFkB-repressing factor, NF-kappaB repressing factor, NFKB repressing factor, NRFNF-kappa-B-repressing factor, Protein ITBA4, Transcription factor NRF
Gene Symbols NKRF
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-296 of human NKRF(NP_060014) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55922
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.