Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.2 Antibody (NX2/2198R), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB385906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385906 Supplier Novus Biologicals Supplier No. NBP308568AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NKX2.2 Monoclonal antibody specifically detects NKX2.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen NKX2.2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NX2/2198R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Homeobox protein NK-2 homolog B, NK-2 (Drosophila) homolog B, NK2 homeobox 2, NK2 transcription factor related, locus 2, NK2 transcription factor related, locus 2 (Drosophila), NK2 transcription factor-like protein B, Nkx2.2, NKX2.2homeobox protein Nkx-2.2, NKX2BNK-2 homolog B
Host Species Rabbit
Immunogen Recombinant human NKX2.2 protein fragment (around aa1-119) (exact sequence is proprietary) (Uniprot: O95096)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4821
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.