Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.8 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB165121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165121 Supplier Novus Biologicals Supplier No. NBP281773AF488
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NKX2.8 Polyclonal antibody specifically detects NKX2.8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NKX2.8
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H
Host Species Rabbit
Immunogen NKX2.8 antibody was raised against a 19 amino acid synthetic peptide near the center of human NKX2.8. The immunogen is located within amino acids 60 - 110 of NKX2.8. Amino Acid Squence: ASPGSDAEKRKKRRVLFSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26257
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.