Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2-4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017071 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017071 Supplier Novus Biologicals Supplier No. NBP281760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKX2-4 Polyclonal antibody specifically detects NKX2-4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen NKX2-4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-20 μg/mL
Formulation PBS
Gene Alias homeobox protein Nkx-2.4, NK2 homeobox 4, NKX2.4, NKX2D
Gene Symbols NKX2-4
Host Species Rabbit
Immunogen NKX2-4 antibody was raised against an 18 amino acid synthetic peptide near the center of human NKX2-4. The immunogen is located within amino acids 220 - 270 of NKX2.8. Amino Acid Squence: KIWFQNHRYKMKRQAKDK
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644524
Test Specificity NKX2.8 antibody is predicted to not cross-react with other NK2 homeobox family members. At least two isoforms of NKX2.8 are known to exist; this antibody will detect both isoforms.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.