Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.8 Antibody (NKX28/2548), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB379370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379370 Supplier Novus Biologicals Supplier No. NBP308638DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen NKX2.8
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone NKX28/2548
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H
Host Species Mouse
Immunogen Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) (Uniprot: O15522)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26257
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.