Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX2.8 Antibody (NKX28/3233R), HRP, Novus Biologicals™
SDP

Catalog No. NB391587 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391587 Supplier Novus Biologicals Supplier No. NBP308653H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NKX2.8 Monoclonal antibody specifically detects NKX2.8 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen NKX2.8
Applications Western Blot, ELISA
Classification Monoclonal
Clone NKX28/3233R
Conjugate HRP
Formulation PBS
Gene Alias Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H
Host Species Rabbit
Immunogen Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) (Uniprot: O15522)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26257
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.