Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3.1 Antibody (0361), FITC, Novus Biologicals™
SDP

Catalog No. NB1001828F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001828F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001828F Supplier Novus Biologicals Supplier No. NB1001828F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKX3.1 Monoclonal specifically detects NKX3.1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen NKX3.1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Western Blot
Classification Monoclonal
Clone 361
Conjugate FITC
Dilution Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. Q99801
Gene Alias BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A
Gene Symbols NKX3-1
Host Species Mouse
Immunogen Full-length human Nkx3.1 produced in bacteria as a hexahistidine-tagged protein. [UniProt# Q99801]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4824
Test Specificity Mouse and Rat. Predicted to work in human based on 100% sequence homology.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.