Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3.1 Antibody (NKX3.1/2576), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB011508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB011508 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB011508 Supplier Novus Biologicals Supplier No. NBP279871JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NKX3.1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NKX3.1/2576
Conjugate Janelia Fluor 646
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A
Gene Symbols NKX3-1
Host Species Mouse
Immunogen Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) (Uniprot: Q99801)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.