Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX6.1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB326987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326987 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326987 Supplier Novus Biologicals Supplier No. NBP299125MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NKX6.1 Polyclonal antibody specifically detects NKX6.1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen NKX6.1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NKX6.1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4825
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.