Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLGN4Y Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7205222 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8780 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8780 Supplier Proteintech Group Inc Supplier No. 134891AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody can recognize both of NLGN4Y and NLGN4X at 72 kDa mature form.

Neuroligins, such as NLGN4Y, are cell adhesion molecules present at the postsynaptic side of the synapse and may be essential for the formation of functional synapses (Jamain et al., 2003 ).

Specifications

Antigen NLGN4Y
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NLGN4Y
Gene Accession No. Q8N0W4, Q8NFZ3
Gene Alias KIAA0951, neuroligin 4, Y linked, Neuroligin Y, NLGN4Y
Gene Symbols NLGN4X, NLGN4Y
Host Species Rabbit
Immunogen NLGN4Y Fusion Protein Ag4311
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22829, 57502
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.