Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLK Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB323523 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323523 Supplier Novus Biologicals Supplier No. NBP299830MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLK Polyclonal antibody specifically detects NLK in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen NLK
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp761G1211, EC 2.7.11, EC 2.7.11.24, FLJ21033, LAK1, nemo like kinase, nemo-like kinaseserine/threonine-protein kinase NLK, Protein LAK1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human NLK (Uniprot#: Q9UBE8; Val121-Glu527)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 51701
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.