Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRC3 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB202132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB202132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB202132 Supplier Novus Biologicals Supplier No. NBP241123V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLRC3 Polyclonal antibody specifically detects NLRC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen NLRC3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human NOD3. The immunogen is located within amino acids 630 - 680 of NOD3. Amino Acid Squence: CRKLRLDTNQFQDPVMEL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 197358
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.