Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRC3 Antibody (Eowyn-1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB173138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173138 Supplier Novus Biologicals Supplier No. NBP280084FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

NLRC3 Monoclonal antibody specifically detects NLRC3 in Mouse samples. It is validated for Western Blot

Specifications

Antigen NLRC3
Applications Western Blot
Classification Monoclonal
Clone Eowyn-1
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3
Host Species Mouse
Immunogen Recombinant mouse NLRC3 (card domain).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 197358
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.