Learn More
Description
Specifications
Specifications
| Antigen | NLRP10/Pynod/NALP10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0-2.0 ug/ml |
| Formulation | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | Q86W26 |
| Gene Alias | CLR11.1, NACHT, leucine rich repeat and PYD containing 10, NALP10Pynod, NLR family, pyrin domain containing 10, NOD8Nucleotide-binding oligomerization domain protein 8, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 10, PAN5, PYNODNACHT, LRR and PYD domains-containing protein 10 |
| Gene Symbols | NLRP10 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
