Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP10/Pynod/NALP10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP185557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP185557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP185557 Supplier Novus Biologicals Supplier No. NBP185557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLRP10/Pynod/NALP10 Polyclonal specifically detects NLRP10/Pynod/NALP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NLRP10/Pynod/NALP10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CLR11.1, NACHT, leucine rich repeat and PYD containing 10, NALP10Pynod, NLR family, pyrin domain containing 10, NOD8Nucleotide-binding oligomerization domain protein 8, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 10, PAN5, PYNODNACHT, LRR and PYD domains-containing protein 10
Gene Symbols NLRP10
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:RLLEVKEQEGNDEMTLTMQFLLDISKKDSFSNLELKFCFRISPCLAQDLKHFKEQMESMKHNRTWDLEFSLYEAKIKNLVKGIQMNNVSFKIKHSNEKKSQ
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338322
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.