Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP13 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176294 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176294 Supplier Novus Biologicals Supplier No. NBP176294
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLRP13 Polyclonal specifically detects NLRP13 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NLRP13
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2.5-20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. EAW72420
Gene Alias CLR19.7, leucine rich repeat and pyrin domain containing 13, NACHT, leucine rich repeat and PYD containing 13, NACHT, LRR and PYD containing protein 13, NACHT, LRR and PYD domains-containing protein 13, NALP13, NLR family, pyrin domain containing 13, NOD14, nucleotide-binding oligomerization domain protein 14, PAN13
Gene Symbols NLRP13
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human NALP13. The immunogen is located within amino acids 150 - 200 of NALP13. Amino Acid Squence: QDPNQEEPEMLEEADHRR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126204
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.