Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP14 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB342552 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342552 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342552 Supplier Novus Biologicals Supplier No. NBP281748PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLRP14 Polyclonal antibody specifically detects NLRP14 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen NLRP14
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias CLR11.2, LRR and PYD containing protein 14, NACHT, leucine rich repeat and PYD containing 14, NACHT, LRR and PYD domains-containing protein 14, NALP14, NLR family, pyrin domain containing 14, nucleotide-binding oligomerization domain protein 5, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 14
Host Species Rabbit
Immunogen NLRP14 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NLRP14. The immunogen is located within amino acids 200 - 250 of NLRP14. Amino Acid Squence: NGREINQLKERSFAQL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 338323
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.