Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP3/NALP3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB170802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170802 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170802 Supplier Novus Biologicals Supplier No. NBP212446AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP3/NALP3 Polyclonal antibody specifically detects NLRP3/NALP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen NLRP3/NALP3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
Host Species Rabbit
Immunogen This NLRP3/NALP3 antibody was raised against a portion of amino acids 1-50 of human NLRP3/NALP3.
Purification Method Immunogen affinity purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114548
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.