Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPRXL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19142320UL
Description
TPRXL Polyclonal specifically detects TPRXL in Human samples. It is validated for Western Blot.Specifications
TPRXL | |
Polyclonal | |
Western Blot 1:1000 | |
TPRXL tetra-peptide repeat homeobox-like | |
Rabbit | |
28 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
TPRXL | |
Synthetic peptide directed towards the C terminal of human TPRXL. Peptide sequence SSSSSSTSSPSTSSPSSSSPSSSSPSSSCPSAALGRRPQSPQSSHCAPFP. | |
Affinity Purified | |
RUO | |
348825 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction