Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPRXL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | TPRXL |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19142320
![]() |
Novus Biologicals
NBP19142320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191423
![]() |
Novus Biologicals
NBP191423 |
100 μL |
Each for $487.50
|
|
|||||
Description
TPRXL Polyclonal specifically detects TPRXL in Human samples. It is validated for Western Blot.Specifications
TPRXL | |
Polyclonal | |
Rabbit | |
Human | |
348825 | |
Synthetic peptide directed towards the C terminal of human TPRXL. Peptide sequence SSSSSSTSSPSTSSPSSSSPSSSSPSSSCPSAALGRRPQSPQSSHCAPFP. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
TPRXL tetra-peptide repeat homeobox-like | |
TPRXL | |
IgG | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title