Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuferricin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19143020UL
Description
Neuferricin Polyclonal specifically detects Neuferricin in Human samples. It is validated for Western Blot.Specifications
Neuferricin | |
Polyclonal | |
Western Blot 1:1000 | |
NP_653212 | |
CYB5D2 | |
Synthetic peptide directed towards the N terminal of human CYB5D2. Peptide sequence RLFIPEELSRYRGGPGDPGLYLALLGRVYDVSSGRRHYEPGSHYSGFAGR. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
cytochrome b5 domain containing 2, cytochrome b5 domain-containing protein 2, MGC32124, neuferricin | |
Rabbit | |
Affinity Purified | |
RUO | |
124936 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction