Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neuferricin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $499.50
Specifications
Antigen | Neuferricin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19143020
![]() |
Novus Biologicals
NBP19143020UL |
20 μL |
Each for $158.00
|
|
|||||
NBP191430
![]() |
Novus Biologicals
NBP191430 |
100 μL |
Each for $499.50
|
|
|||||
Description
Neuferricin Polyclonal specifically detects Neuferricin in Human samples. It is validated for Western Blot.Specifications
Neuferricin | |
Polyclonal | |
Rabbit | |
NP_653212 | |
124936 | |
Synthetic peptide directed towards the N terminal of human CYB5D2. Peptide sequence RLFIPEELSRYRGGPGDPGLYLALLGRVYDVSSGRRHYEPGSHYSGFAGR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
cytochrome b5 domain containing 2, cytochrome b5 domain-containing protein 2, MGC32124, neuferricin | |
CYB5D2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title