Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | MCM10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18239520
![]() |
Novus Biologicals
NBP18239520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP182395
![]() |
Novus Biologicals
NBP182395 |
100 μL |
Each for $487.50
|
|
|||||
Description
MCM10 Polyclonal specifically detects MCM10 in Mouse samples. It is validated for Western Blot.Specifications
MCM10 | |
Polyclonal | |
Rabbit | |
NP_081566 | |
55388 | |
Synthetic peptide towards Mcm10. Peptide sequence DEFDELFDADGDGESYTEEAGSGEEGKTGNQEERLATLFGDVEDLTDDEV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CNA43homolog of yeast MCM10, DNA43, hsMCM10, MCM10 minichromosome maintenance deficient 10, MCM10 minichromosome maintenance deficient 10 (S. cerevisiae), MGC126776, minichromosome maintenance complex component 10, PRO2249, protein MCM10 homolog | |
MCM10 | |
IgG | |
98 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title