Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C11orf16 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19148820UL

 View more versions of this product

Catalog No. NBP19148820


Only null left
Add to Cart

Description

Description

C11orf16 Polyclonal specifically detects C11orf16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C11orf16
Polyclonal
Western Blot 1:1000
NP_065694
C11ORF16
Synthetic peptide directed towards the middle region of human C11orf16. Peptide sequence EPCLGKPGTRYSNICKEEKDHKQQRAQTAVVGTTKELVSKATHMKPPRTP.
Affinity Purified
RUO
56673
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
chromosome 11 open reading frame 16, hypothetical protein LOC56673
Rabbit
51 kDa
20 μL
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.