Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C11orf16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | C11orf16 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19148820
![]() |
Novus Biologicals
NBP19148820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191488
![]() |
Novus Biologicals
NBP191488 |
100 μL |
Each for $487.50
|
|
|||||
Description
C11orf16 Polyclonal specifically detects C11orf16 in Human samples. It is validated for Western Blot.Specifications
C11orf16 | |
Polyclonal | |
Rabbit | |
Human | |
chromosome 11 open reading frame 16, hypothetical protein LOC56673 | |
C11ORF16 | |
IgG | |
51 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_065694 | |
56673 | |
Synthetic peptide directed towards the middle region of human C11orf16. Peptide sequence EPCLGKPGTRYSNICKEEKDHKQQRAQTAVVGTTKELVSKATHMKPPRTP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title