Learn More
Description
Specifications
Specifications
| Antigen | Melanoma antigen family E1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | NP_444431 |
| Gene Alias | Alpha-dystrobrevin-associated MAGE Protein, DAMAGEMAGE-E1 antigen, HCA1, hepatocellular carcinoma-associated HCA1, Hepatocellular carcinoma-associated protein 1, KIAA1587dystrobrevin-associated MAGE protein, melanoma antigen family E, 1, melanoma-associated antigen E1 |
| Gene Symbols | MAGEE1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
