Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OTUD6A Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19149820UL

 View more versions of this product

Catalog No. NBP19149820


Only null left
Add to Cart

Description

Description

OTUD6A Polyclonal specifically detects OTUD6A in Human samples. It is validated for Western Blot.
Specifications

Specifications

OTUD6A
Polyclonal
Western Blot 1:1000
NP_997203
OTUD6A
Synthetic peptide directed towards the middle region of human OTUD6A. Peptide sequence KMNLENRPPRSSKAHRKRERMESEERERQESIFQAEMSEHLAGFKREEEE.
Affinity Purified
RUO
139562
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
DUBA2, DUBA-2, OTU domain containing 6A
Rabbit
32 kDa
20 μL
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.