Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prodynorphin/PDYN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19150020UL
Description
Dynorphin Polyclonal specifically detects Dynorphin in Human, Mouse samples. It is validated for Western Blot.Specifications
PDYN | |
Polyclonal | |
Western Blot 1:1000 | |
NP_061351 | |
PDYN | |
Synthetic peptide directed towards the middle region of human Pdyn. Peptide sequence TLTVSGLRGKDDLEDEVALEEGYSALAKLLEPVLKELEKSRLLTSVPEEK. | |
Affinity Purified | |
RUO | |
5173 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ADCA, Beta-neoendorphin-dynorphin, leu-enkephalin, leumorphin, MGC26418, PENKB, preprodynorphin, preproenkephalin B, prodynorphin, proenkephalin-B, rimorphin, SCA23, spinocerebellar ataxia 23 | |
Rabbit | |
28 kDa | |
20 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction