Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prodynorphin/PDYN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Applications | Western Blot |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19150020
![]() |
Novus Biologicals
NBP19150020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191500
![]() |
Novus Biologicals
NBP191500 |
100 μL |
Each for $487.50
|
|
|||||
Description
Prodynorphin/PDYN Polyclonal specifically detects Prodynorphin/PDYN in Human samples. It is validated for Western Blot.Specifications
Western Blot | |
Unconjugated | |
RUO | |
PDYN | |
Primary |
Polyclonal | |
Rabbit | |
5173 | |
IgG | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title