Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    C9orf64 Antibody, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191505
Description
C9orf64 Polyclonal specifically detects C9orf64 in Human samples. It is validated for Western Blot.Specifications
| C9orf64 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| chromosome 9 open reading frame 64, hypothetical protein LOC84267, RP11-575L7.5 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 79%. | |
| Human, Bovine, Canine | |
| IgG | 
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_115683 | |
| C9ORF64 | |
| Synthetic peptide directed towards the C terminal of human C9orf64. Peptide sequence EMLSYGDRQEVEIRGCSLWCVELIRDCLLELIEQKGEKPNGEINSILLDY. | |
| Affinity purified | |
| RUO | |
| 84267 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            