Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf64 Antibody, Novus Biologicals™
SDP

Catalog No. NBP19150620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19150620 20 μL
NBP191506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19150620 Supplier Novus Biologicals Supplier No. NBP19150620UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

C9orf64 Polyclonal specifically detects C9orf64 in Human samples. It is validated for Western Blot.

Specifications

Antigen C9orf64
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_115683
Gene Alias chromosome 9 open reading frame 64, hypothetical protein LOC84267, RP11-575L7.5
Gene Symbols C9orf64
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human C9orf64. Peptide sequence RVEGWKALHELNPRAADEAAVNWVFVTDTLNFSFWSEQDEHKCVVRYRGK.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84267
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.