Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCF23 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP19153320UL

 View more versions of this product

Catalog No. NBP19153320


Description

Description

TCF23 Polyclonal specifically detects TCF23 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TCF23
Polyclonal
Western Blot 1:1000
NP_444315
TCF23
Synthetic peptide directed towards the C terminal of mouse TCF23. Peptide sequence PMRSRLYAGGLGCSDLDSTTAITTGQRCKDAELGSQDSVAAESLLTSPAF.
Protein A purified
RUO
150921
Store at -20C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
BHLHA24, class A basic helix-loop-helix protein 24, class II basic helix-loop-helix protein TCF23, TCF-23, transcription factor 23
Rabbit
23 kDa
20 μL
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.